Learn More
Bachem Z-Arg-Leu-Arg-Gly-Gly-AMC
SDP
Catalog No. 50260344
Encompass_Preferred
Z-Arg-Leu-Arg-Gly-Gly-AMC Acetate, 167698-69-3, C40H56N12O9, Mr 848.96, 100mg. Z-RLRGG-AMC is a fluorogenic substrate for the deubiquitinating enzyme isopeptidase T (IPaseT) and other ubiquitin C-terminal hydrolases (UCHs) based on the C-termini of ubiquitin with a kcat/Km value of 95 M-1s-1. Together with our product I-1685 it is used in continuous assays for precise mechanistic studies or microtiter plate assays for high-throughput inhibitor screening.
Catalog No. 50-260-344 Supplier Bachem Supplier No. 4027158.0100
Z-Arg-Leu-Arg-Gly-Gly-AMC Acetate, 167698-69-3, C40H56N12O9, Mr 848.96, 100mg. Z-RLRGG-AMC is a fluorogenic substrate for the deubiquitinating enzyme isopeptidase T (IPaseT) and other ubiquitin C-terminal hydrolases (UCHs) based on the C-termini of ubiquitin with a kcat/Km value of 95 M-1s-1. Together with our product I-1685 it is used in continuous assays for precise mechanistic studies or microtiter plate assays for high-throughput inhibitor screening.