Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Z-Arg-SBzl | 88253-86-5 | MFCD00222045 | 96.58% | 414.52 | 10 MG
SDP

Catalog No. 5000398481
Encompass_Preferred

Z-Arg-SBzl is a substrate of both bovine and activated human protein C.

  • CAS number: 88253-86-5
  • Molecular weight: 414.52
  • Formula: C21H26N4O3S
  • Appearance: Solid, white to off-white
  • Functions as a substrate for bovine and activated human protein C

Catalog No. 50-003-98481 Supplier Medchemexpress LLC Supplier No. HYP512510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Z-Arg-SBzl is a substrate of both bovine and activated human protein C.

  • CAS number: 88253-86-5
  • Molecular weight: 414.52
  • Formula: C21H26N4O3S
  • Appearance: Solid, white to off-white
  • Functions as a substrate for bovine and activated human protein C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.