Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Z-Asn-ONp, Advanced ChemTech

Catalog No. 01338259
Click to view available options
Quantity:
1 g
5 g
25 g

Catalog No. 01-338-259

Supplier: Advanced Chem Tech ZN21511G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3256-57-3
Molecular Formula C18H17N3O7
Molecular Weight (g/mol) 387.35
MDL Number MFCD00024661
InChI Key YLWIFNIVONXXMG-OAHLLOKOSA-N
IUPAC Name 4-nitrophenyl (2R)-2-{[(benzyloxy)carbonyl]amino}-3-carbamoylpropanoate
SMILES NC(=O)C[C@@H](NC(=O)OCC1=CC=CC=C1)C(=O)OC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Color White to Off-White
Quantity 1 g
Grade Biochemical
Chemical Name or Material Z-Asn-ONp
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.