Learn More
Medchemexpress LLC HY-113953 5mg Medchemexpress, Z-Asp-CH2-DCB CAS:153088-73-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1139535MG
Encompass_Preferred
Medchemexpress, HY-113953 5mg Z-Asp-CH2-DCB CAS:153088-73-4 Z-Asp-CH2-DCB is an irreversible broad spectrum caspase inhibitor. Z-Asp-CH2-DCB also inhibits proteases with caspase-like activity. Z-D-CH2-DCB blocks the production of IL-1β, TNF-α, IL-6, and IFN-γ in staphylococcal enterotoxin B (SEB)-stimulated peripheral blood mononuclear cells (PBMC), and reduces SEB-1-stimulated T-cell proliferation in a dose-dependent manner. Z-Asp-CH2-DCB prevents SU5416-induced septal cell apoptosis and emphysema development[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9269