Learn More
Abcam Z-Asp-CH2-DCB, caspase inhibitor, 10MG
Catalog No. 502705572
Encompass_Preferred
MW 454.3 Da, Purity >98%. Irreversible, cell permeable broad-spectrum caspase inhibitor (IC₅₀ values are 1.1 and 4.1 μM for caspase-3 and caspase-6 respectively). Inhibits hypoxia-induced apoptosis. Shows anti-ischemic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5572 Supplier Abcam Supplier No. AB14460710MG
MW 454.3 Da, Purity >98%. Irreversible, cell permeable broad-spectrum caspase inhibitor (IC₅₀ values are 1.1 and 4.1 μM for caspase-3 and caspase-6 respectively). Inhibits hypoxia-induced apoptosis. Shows anti-ischemic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement