Learn More
CPC Scientific Z-Ala-Thr-Ala-Asp-FMK 0.5MG
SDP
Catalog No. 502109080
Encompass
SEQUENCE: Z-Ala-Thr-Ala-Asp-FMKONE-LETTER SEQUENCE: Z-ATAD-FMKMOLECULAR FORMULA: C23H31F1N4O9MOLECULAR WEIGHT: 526.5STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: ApoptosisREFERENCES:SKU(s): CASP-086A, CASP-086B
Catalog No. 50-210-9080 Supplier CPC Scientific Supplier No. CASP086A
SEQUENCE: Z-Ala-Thr-Ala-Asp-FMKONE-LETTER SEQUENCE: Z-ATAD-FMKMOLECULAR FORMULA: C23H31F1N4O9MOLECULAR WEIGHT: 526.5STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: ApoptosisREFERENCES:SKU(s): CASP-086A, CASP-086B