Learn More
Abcam Z-ATAD-FMK, Caspase-12 inhibitor, 5MG
Catalog No. 502706655
Encompass_Preferred
MW 540.54 g/mol, Purity >95%. Specific caspase-12 inhibitor. Suppresses annular cell apoptosis and caspase-9 activity in vitro. Active in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6655 Supplier Abcam Supplier No. AB1413835MG
MW 540.54 g/mol, Purity >95%. Specific caspase-12 inhibitor. Suppresses annular cell apoptosis and caspase-9 activity in vitro. Active in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement