Learn More
World Precision Instrument Z-Axis Vertical Extension for Quad Manipulator
SDP
Catalog No. 502539227
Encompass
  • For use with the SU-QUAD when the diagonal axis is too tall
  • For use with the SU-QUAD when the angle of approach is too steep
Catalog No. 50-253-9227 Supplier World Precision Instrument Supplier No. 505133
  • For use with the SU-QUAD when the diagonal axis is too tall
  • For use with the SU-QUAD when the angle of approach is too steep