Learn More
Z BIOTECH LLC Z BIOTECH LLC Sialoside Array Slide, 16-sample, 50 Sialoside Structures
SDP
Catalog No. NC3977888
Encompass
The Sialoside Array is designed for researchers to investigate interactions between sialosides and various biological samples, including proteins, antibodies, and cells. It features 50 distinct N-glycans and LacNAc glycans, each paired with Neu5Gc and Neu5Ac. This platform offers high-throughput, reliable sialoside-binding information with a simple assay format, requiring small sample volumes and yielding results quickly. Arrays are available in 8 or 16 identical subarray formats for simultaneous multi-sample analysis.
  • 50 Sialoside structures
  • Unrivaled sensitivity and specificity
  • Simple assay format
  • Small sample volume
  • Customizable (select glycans for a specific microarray format)
  • Assay service available
Catalog No. NC3977888 Supplier Z BIOTECH LLC Supplier No. 1060616S
The Sialoside Array is designed for researchers to investigate interactions between sialosides and various biological samples, including proteins, antibodies, and cells. It features 50 distinct N-glycans and LacNAc glycans, each paired with Neu5Gc and Neu5Ac. This platform offers high-throughput, reliable sialoside-binding information with a simple assay format, requiring small sample volumes and yielding results quickly. Arrays are available in 8 or 16 identical subarray formats for simultaneous multi-sample analysis.
  • 50 Sialoside structures
  • Unrivaled sensitivity and specificity
  • Simple assay format
  • Small sample volume
  • Customizable (select glycans for a specific microarray format)
  • Assay service available