Learn More
Cayman Chemical (Z)-Capsaicin, Purity: ≥95%, CAS: 25775-90-0, 10 mg
SDP
Catalog No. 502978546
Encompass_Preferred
An alkaloid with diverse biological activities; an agonist of TRPV1 (EC50 = 28.2 nM for inducing calcium uptake in CHO cells expressing human TRPV1); shortens the action potential duration in isolated canine Purkinje fibers at 10 µM; increases the latency to paw withdrawal in the hot plate test in rats at 0.68 mmol/animal; topical application reduces croton oil-induced ear edema in mice; induces emesis in musk shrews; decreases disease severity in a guinea pig model of HSV-2-induced genital herpes
Catalog No. 50-297-8546 Supplier Cayman Chemical Supplier No. 3992910 MG
An alkaloid with diverse biological activities; an agonist of TRPV1 (EC50 = 28.2 nM for inducing calcium uptake in CHO cells expressing human TRPV1); shortens the action potential duration in isolated canine Purkinje fibers at 10 µM; increases the latency to paw withdrawal in the hot plate test in rats at 0.68 mmol/animal; topical application reduces croton oil-induced ear edema in mice; induces emesis in musk shrews; decreases disease severity in a guinea pig model of HSV-2-induced genital herpes