Learn More
Medchemexpress LLC HY-B1583 10mg Medchemexpress, (Z)-Capsaicin CAS:25775-90-0 Purity:>98%
SDP
Catalog No. 501932972
Encompass_Preferred
Medchemexpress, HY-B1583 10mg (Z)-Capsaicin CAS:25775-90-0 (Z)-Capsaicin is the cis isomer of capsaicin, acts as an orally active TRPV1 agonist, and is used in the research of neuropathic pain. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2972 Supplier Medchemexpress LLC Supplier No. HYB158310MG
Medchemexpress, HY-B1583 10mg (Z)-Capsaicin CAS:25775-90-0 (Z)-Capsaicin is the cis isomer of capsaicin, acts as an orally active TRPV1 agonist, and is used in the research of neuropathic pain. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.