Learn More
Medchemexpress LLC (Z)-Capsaicin | 25775-90-0 | 99.79% | 1 ML
SDP
Catalog No. 5000412740
Encompass_Preferred
(Z)-Capsaicin is the cis isomer of capsaicin, acts as an orally active TRPV1 agonist, and is used in the research of neuropathic pain. It is for research use only and not sold to patients.
  • Cis isomer of capsaicin.
  • Orally active TRPV1 agonist.
  • Used in the research of neuropathic pain.
  • May also block neuronal calcium channels.
  • Significantly attenuates nociceptive behavior after oral administration in rats.
  • Tolerated in primary or recurrent experimental genital herpes simplex virus infection in guinea pigs.
Catalog No. 50-004-12740 Supplier Medchemexpress LLC Supplier No. HYB158310MM/1ML
(Z)-Capsaicin is the cis isomer of capsaicin, acts as an orally active TRPV1 agonist, and is used in the research of neuropathic pain. It is for research use only and not sold to patients.
  • Cis isomer of capsaicin.
  • Orally active TRPV1 agonist.
  • Used in the research of neuropathic pain.
  • May also block neuronal calcium channels.
  • Significantly attenuates nociceptive behavior after oral administration in rats.
  • Tolerated in primary or recurrent experimental genital herpes simplex virus infection in guinea pigs.