Learn More
Zymo Research Corporation Mix & Go E. coli Transformation Kit & Buffer Set, up to 20 ml
SDP
Catalog No. NC9756453
Encompass_Preferred
The methods eliminate the requirement of heat-shocking and related procedures. Instead, transformation can be performed by adding DNA to prepared Mix & Go cells and the mixture spread directly to a culture plate. Transformation efficiencies typically range from 108–109 transformants/µg of pUC19 DNA but can vary depending on the strain of E. coli. Most E. coli strains respond well to the Mix & Go preparation method and demonstrate fast transformation kinetics.The procedures are easy. Simply culture the E. coli strain of your choice in ZymoBroth™ medium (or SOB), wash and then resuspend the cells in the provided uniquely formulated buffers. The cells are now ready for transformation.
Catalog No. NC9756453 Supplier Zymo Research Corporation Supplier No. T3001
The methods eliminate the requirement of heat-shocking and related procedures. Instead, transformation can be performed by adding DNA to prepared Mix & Go cells and the mixture spread directly to a culture plate. Transformation efficiencies typically range from 108–109 transformants/µg of pUC19 DNA but can vary depending on the strain of E. coli. Most E. coli strains respond well to the Mix & Go preparation method and demonstrate fast transformation kinetics.The procedures are easy. Simply culture the E. coli strain of your choice in ZymoBroth™ medium (or SOB), wash and then resuspend the cells in the provided uniquely formulated buffers. The cells are now ready for transformation.