Learn More
Medchemexpress LLC Z-D-Lys-OH | 70671-54-4 | 99.9% | 280.32 | 25 G
SDP
Catalog No. 5000451464
Encompass_Preferred
Z-D-Lys-OH, also known as N-benzoxycarbonyl-D-lysine or Nα-Cbz-D-lysine, is a white crystalline organic compound with the molecular formula C14H20N2O4 and a molecular weight of 280.32. It is identified by CAS No. 70671-54-4.
  • Precise chemical identity with well-defined structure.
  • Specific stereoisomeric form, crucial for chirality-sensitive applications.
  • Potential for various interactions in chemical and biological systems due to hydrogen bond donor and acceptor counts.
  • Comprehensive identification with multiple synonyms for easy cross-referencing.
  • Detailed IR and Raman spectral information available for characterization and quality control.
  • Suitable for peptide synthesis.
  • Soluble in common organic solvents like chloroform, dichloromethane, ethyl acetate, DMSO, and acetone.
Catalog No. 50-004-51464 Supplier Medchemexpress LLC Supplier No. 25GHYW013297
Z-D-Lys-OH, also known as N-benzoxycarbonyl-D-lysine or Nα-Cbz-D-lysine, is a white crystalline organic compound with the molecular formula C14H20N2O4 and a molecular weight of 280.32. It is identified by CAS No. 70671-54-4.
  • Precise chemical identity with well-defined structure.
  • Specific stereoisomeric form, crucial for chirality-sensitive applications.
  • Potential for various interactions in chemical and biological systems due to hydrogen bond donor and acceptor counts.
  • Comprehensive identification with multiple synonyms for easy cross-referencing.
  • Detailed IR and Raman spectral information available for characterization and quality control.
  • Suitable for peptide synthesis.
  • Soluble in common organic solvents like chloroform, dichloromethane, ethyl acetate, DMSO, and acetone.