Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Z-D-Lys-OH | 70671-54-4 | 99.96% | 280.32 | 10 G
SDP

Catalog No. 5000454316
Encompass_Preferred

Z-D-Lys-OH is a laboratory chemical intended for research use only. It is a white to off-white solid with a molecular weight of 280.32 and a purity of 99.96% (HPLC). It should be stored at 4°C, protected from light.

  • CAS number: 70671-54-4
  • Molecular formula: C₁₄H₂₀N₂O₄
  • Molecular weight: 280.32
  • Appearance: White to off-white solid
  • Purity: 99.96% (HPLC)
  • Storage conditions: 4°C, protect from light
  • Identified uses: Laboratory chemicals, manufacture of substances
  • For research use only

Catalog No. 50-004-54316 Supplier Medchemexpress LLC Supplier No. HYW01329710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Z-D-Lys-OH is a laboratory chemical intended for research use only. It is a white to off-white solid with a molecular weight of 280.32 and a purity of 99.96% (HPLC). It should be stored at 4°C, protected from light.

  • CAS number: 70671-54-4
  • Molecular formula: C₁₄H₂₀N₂O₄
  • Molecular weight: 280.32
  • Appearance: White to off-white solid
  • Purity: 99.96% (HPLC)
  • Storage conditions: 4°C, protect from light
  • Identified uses: Laboratory chemicals, manufacture of substances
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.