Learn More
Enzo Life Sciences Z-D(OMe)E(OMe)VD(OMe)-FMK (1mg)
SDP
Catalog No. 502008284
Encompass_Preferred
Irreversible, cell permeable inhibitor of caspase-3 and -7. Alternative name: Caspase 3 inhibitor (fluoromethylketone), Caspase 7 inhibitor (fluoromethylketone). Formula: C30H41FN4O12. MW: 668.7. Purity: ≥95% (TLC). Formulation: Lyophilized. Solubility: Soluble in DMSO (20mM) or dimethyl formamide. Long Term Storage: -20°C.
Catalog No. 50-200-8284 Supplier Enzo Life Sciences Supplier No. ALX260072M001
Irreversible, cell permeable inhibitor of caspase-3 and -7. Alternative name: Caspase 3 inhibitor (fluoromethylketone), Caspase 7 inhibitor (fluoromethylketone). Formula: C30H41FN4O12. MW: 668.7. Purity: ≥95% (TLC). Formulation: Lyophilized. Solubility: Soluble in DMSO (20mM) or dimethyl formamide. Long Term Storage: -20°C.