Learn More
Medchemexpress LLC Z-D-Phg-OH (D-Cbz phenylglycine) | 17609-52-8 | ≥97.0% | C16H15NO4 | 500 G
SDP
Catalog No. 5000434546
Encompass_Preferred
Z-D-Phg-OH (D-Cbz phenylglycine) is an N-blocked amino acid with Kd values of 390 μM and 323 μM for tBuCQN and tBuCQD, respectively. It is for research use only.
  • Peptides
  • Reagents for peptide synthesis
  • Amino acids and derivatives
  • Others amino acid derivatives
Catalog No. 50-004-34546 Supplier Medchemexpress LLC Supplier No. HY75332500G
Z-D-Phg-OH (D-Cbz phenylglycine) is an N-blocked amino acid with Kd values of 390 μM and 323 μM for tBuCQN and tBuCQD, respectively. It is for research use only.
  • Peptides
  • Reagents for peptide synthesis
  • Amino acids and derivatives
  • Others amino acid derivatives