Learn More
Medchemexpress LLC Z-D-Phg-OH (D-Cbz phenylglycine) | 17609-52-8 | MFCD00021703 | ≥97.0% | C16H15NO4 | 100 G
SDP
Catalog No. 5000433599
Encompass_Preferred
Z-D-Phg-OH (D-Cbz phenylglycine) is an N-blocked amino acid. It is used in research and exhibits Kd values of 390 μM for tBuCQN and 323 μM for tBuCQD.
  • Classified as an N-blocked amino acid
  • Exhibits Kd values of 390 μM for tBuCQN
  • Exhibits Kd values of 323 μM for tBuCQD
  • Intended for research use only
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-004-33599 Supplier Medchemexpress LLC Supplier No. HY75332100G
Z-D-Phg-OH (D-Cbz phenylglycine) is an N-blocked amino acid. It is used in research and exhibits Kd values of 390 μM for tBuCQN and 323 μM for tBuCQD.
  • Classified as an N-blocked amino acid
  • Exhibits Kd values of 390 μM for tBuCQN
  • Exhibits Kd values of 323 μM for tBuCQD
  • Intended for research use only
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month