Learn More
Medchemexpress LLC Z-D-Ser-OH | 6081-61-4 | 565770 | 99.5% | 239.22 | 25 G
SDP
Catalog No. 5000453299
Encompass_Preferred
Z-D-Ser-OH is a serine derivative used for research purposes. Amino acids and their derivatives are commercially utilized as ergogenic supplements, influencing the secretion of anabolic hormones, fuel supply during exercise, mental performance under stress, and preventing exercise-induced muscle damage. They are recognized for their benefits as ergogenic dietary substances.
  • Serine derivative
  • Used for research only
  • Can be used in custom peptide synthesis
  • Amino acid derivatives can influence anabolic hormone secretion, fuel supply during exercise, and mental performance
  • May prevent exercise-induced muscle damage
  • Ships at room temperature in the continental US
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-004-53299 Supplier Medchemexpress LLC Supplier No. 25GHYW012104
Z-D-Ser-OH is a serine derivative used for research purposes. Amino acids and their derivatives are commercially utilized as ergogenic supplements, influencing the secretion of anabolic hormones, fuel supply during exercise, mental performance under stress, and preventing exercise-induced muscle damage. They are recognized for their benefits as ergogenic dietary substances.
  • Serine derivative
  • Used for research only
  • Can be used in custom peptide synthesis
  • Amino acid derivatives can influence anabolic hormone secretion, fuel supply during exercise, and mental performance
  • May prevent exercise-induced muscle damage
  • Ships at room temperature in the continental US
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month