Learn More
Medchemexpress LLC Z-D-Ser(TBu)-OH | 65806-90-8 | 295.33 | 100 G
SDP
Catalog No. 5000454994
Encompass_Preferred
Z-D-Ser(TBu)-OH is a serine derivative. Amino acids and amino acid derivatives, such as Z-D-Ser(TBu)-OH, are commercially used as ergogenic supplements. They impact the secretion of anabolic hormones, fuel supply during exercise, mental performance in stress-related tasks, and help prevent exercise-induced muscle damage. They are recognized as beneficial ergogenic dietary substances.
  • Influence the secretion of anabolic hormones.
  • Supply fuel during exercise.
  • Improve mental performance in stress-related tasks.
  • Help prevent exercise-induced muscle damage.
Catalog No. 50-004-54994 Supplier Medchemexpress LLC Supplier No. HYW011444100G
Z-D-Ser(TBu)-OH is a serine derivative. Amino acids and amino acid derivatives, such as Z-D-Ser(TBu)-OH, are commercially used as ergogenic supplements. They impact the secretion of anabolic hormones, fuel supply during exercise, mental performance in stress-related tasks, and help prevent exercise-induced muscle damage. They are recognized as beneficial ergogenic dietary substances.
  • Influence the secretion of anabolic hormones.
  • Supply fuel during exercise.
  • Improve mental performance in stress-related tasks.
  • Help prevent exercise-induced muscle damage.