Learn More
Medchemexpress LLC Z-DEVD-AFC | 1135416-11-3 | 98.1% | 821.71 | 10 MG
SDP
Catalog No. 5000423968
Encompass_Preferred
Z-DEVD-AFC is a cell-permeant substrate for caspase-3, which causes a shift in fluorescence upon cleavage of the AFC fluorophore.
  • Can be used to detect caspase-3-like enzyme activity.
  • For research use only.
Catalog No. 50-004-23968 Supplier Medchemexpress LLC Supplier No. HYP198610MG
Z-DEVD-AFC is a cell-permeant substrate for caspase-3, which causes a shift in fluorescence upon cleavage of the AFC fluorophore.
  • Can be used to detect caspase-3-like enzyme activity.
  • For research use only.