Learn More
Medchemexpress LLC Z-DEVD-AFC | 1135416-11-3 | 98.1% | 821.71 | 5 MG
SDP
Catalog No. 5000422265
Encompass_Preferred
Z-DEVD-AFC is a cell-permeant substrate for caspase-3. It causes a shift in fluorescence upon cleavage of the AFC fluorophore, allowing for detection of caspase-3-like enzyme activity. This product is for research use only.
  • Cell-permeant substrate for caspase-3
  • Fluorescence shift upon AFC fluorophore cleavage
  • Detects caspase-3-like enzyme activity
  • White to off-white solid
  • Emission (Em): 500
  • Excitation (Ex): 380
Catalog No. 50-004-22265 Supplier Medchemexpress LLC Supplier No. HYP19865MG
Z-DEVD-AFC is a cell-permeant substrate for caspase-3. It causes a shift in fluorescence upon cleavage of the AFC fluorophore, allowing for detection of caspase-3-like enzyme activity. This product is for research use only.
  • Cell-permeant substrate for caspase-3
  • Fluorescence shift upon AFC fluorophore cleavage
  • Detects caspase-3-like enzyme activity
  • White to off-white solid
  • Emission (Em): 500
  • Excitation (Ex): 380