Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Z-DEVD-AMC | 98.2% | 767.74 | 10 MG
SDP

Catalog No. 5000398278
Encompass_Preferred

Z-DEVD-AMC is a selective caspase-3 substrate that can be measured by fluorescence spectrometry. It serves as a fluorescence reference standard for AMC-based enzyme substrates, including AMC-based caspase substrates.

  • Selective caspase-3 substrate
  • Measured by fluorescence spectrometry
  • Functions as a fluorescence reference standard for AMC-based enzyme substrates
  • Purity of 98.2%
  • Molecular weight of 767.74
  • White to off-white solid appearance
  • Research use only
  • Excitation wavelength of 390
  • Emission wavelength of 480

Catalog No. 50-003-98278 Supplier Medchemexpress LLC Supplier No. HYP336310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Z-DEVD-AMC is a selective caspase-3 substrate that can be measured by fluorescence spectrometry. It serves as a fluorescence reference standard for AMC-based enzyme substrates, including AMC-based caspase substrates.

  • Selective caspase-3 substrate
  • Measured by fluorescence spectrometry
  • Functions as a fluorescence reference standard for AMC-based enzyme substrates
  • Purity of 98.2%
  • Molecular weight of 767.74
  • White to off-white solid appearance
  • Research use only
  • Excitation wavelength of 390
  • Emission wavelength of 480

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.