Learn More
Medchemexpress LLC Z-DEVD-AMC | 98.2% | 767.74 | 5 MG
SDP
Catalog No. 5000396925
Encompass_Preferred
Z-DEVD-AMC is a selective caspase-3 substrate that can be measured by fluorescence spectrometry. AMC (7-amino-4-methylcoumarin) can be used as a fluorescence reference standard for AMC-based enzyme substrates, including AMC-based caspase substrates. It is for research use only.
  • Selective caspase-3 substrate
  • Can be measured by fluorescence spectrometry
  • AMC can be used as a fluorescence reference standard for AMC-based enzyme substrates
  • For research use only
Catalog No. 50-003-96925 Supplier Medchemexpress LLC Supplier No. HYP33635MG
Z-DEVD-AMC is a selective caspase-3 substrate that can be measured by fluorescence spectrometry. AMC (7-amino-4-methylcoumarin) can be used as a fluorescence reference standard for AMC-based enzyme substrates, including AMC-based caspase substrates. It is for research use only.
  • Selective caspase-3 substrate
  • Can be measured by fluorescence spectrometry
  • AMC can be used as a fluorescence reference standard for AMC-based enzyme substrates
  • For research use only