Learn More
CPC Scientific Z-Asp-Glu-Val-Asp-CMK 1MG
SDP
Catalog No. 502109031
Encompass
ONE-LETTER SEQUENCE: Z-DEVD-CMKMOLECULAR FORMULA: C27H35Cl1N4O12MOLECULAR WEIGHT:643STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [250584-13-5], RESEARCH AREA: ApoptosisREFERENCES: R. Schäfer et al., Exp. Cell Res. , 240, 28 (1998); S. Bourteele et al., J. Biol. Chem., 273, 13245 (1998)
Catalog No. 50-210-9031 Supplier CPC Scientific Supplier No. CASP063A
ONE-LETTER SEQUENCE: Z-DEVD-CMKMOLECULAR FORMULA: C27H35Cl1N4O12MOLECULAR WEIGHT:643STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [250584-13-5], RESEARCH AREA: ApoptosisREFERENCES: R. Schäfer et al., Exp. Cell Res. , 240, 28 (1998); S. Bourteele et al., J. Biol. Chem., 273, 13245 (1998)