Learn More
Medchemexpress LLC HY-12466 1mg Medchemexpress, Z-DEVD-FMK CAS:210344-95-9 Purity:>98%
SDP
Catalog No. 502028881
Encompass_Preferred
Medchemexpress, HY-12466 1mg Z-DEVD-FMK CAS:210344-95-9 Z-DEVD-FMK is a specific and irreversible caspase-3 inhibitor with an IC50 of 18 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8881 Supplier Medchemexpress LLC Supplier No. HY124661MG
Medchemexpress, HY-12466 1mg Z-DEVD-FMK CAS:210344-95-9 Z-DEVD-FMK is a specific and irreversible caspase-3 inhibitor with an IC50 of 18 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.