Learn More
TARGETMOL CHEMICALS INC Z-DEVD-FMK 5MG
SDP
Catalog No. 502579690
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Z-DEVD-FMK (Caspase-3 Inhibitor) is a selective irreversible Caspase-3 inhibitor and also exhibits effective inhibition activity on caspase-6 caspase-7 caspase-8 and caspase-10. purity: 99%
Catalog No. 50-257-9690 Supplier TARGETMOL CHEMICALS INC Supplier No. T60055MG
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Z-DEVD-FMK (Caspase-3 Inhibitor) is a selective irreversible Caspase-3 inhibitor and also exhibits effective inhibition activity on caspase-6 caspase-7 caspase-8 and caspase-10. purity: 99%