Learn More
Medchemexpress LLC HY-12466 10mg , Z-DEVD-FMK Caspase-3 Inhibitor CAS:210344-95-9 Purity:98%
SDP
Catalog No. 501494059
Encompass_Preferred
Medchemexpress, HY-12466 10mg Z-DEVD-FMK Caspase-3 Inhibitor CAS:210344-95-9 Formula:C30H41FN4O12 IC50: 18 μM (caspase-3) Purity:98% Z-DEVD-FMK is a specific and irreversible caspase-3 inhibitor with IC 50 of 18 μM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-4059 Supplier Medchemexpress LLC Supplier No. HY1246610MG
Medchemexpress, HY-12466 10mg Z-DEVD-FMK Caspase-3 Inhibitor CAS:210344-95-9 Formula:C30H41FN4O12 IC50: 18 μM (caspase-3) Purity:98% Z-DEVD-FMK is a specific and irreversible caspase-3 inhibitor with IC 50 of 18 μM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.