Learn More
Enzo Life Sciences Z-DEVD-FMK (Ready-to-Use) (20µl)
SDP
Catalog No. 502008346
Encompass_Preferred
Inhibitor of caspase-3 and -7. Irreversible, cell permeable inhibitor of caspase-3 and -7. Alternative name: Caspase 3 inhibitor (fluoromethylketone), Caspase 7 inhibitor (fluoromethylketone). MW: 668.0. Purity: ≥98% (HPLC). Formulation: Liquid. In DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8346 Supplier Enzo Life Sciences Supplier No. ALX260141R020
Inhibitor of caspase-3 and -7. Irreversible, cell permeable inhibitor of caspase-3 and -7. Alternative name: Caspase 3 inhibitor (fluoromethylketone), Caspase 7 inhibitor (fluoromethylketone). MW: 668.0. Purity: ≥98% (HPLC). Formulation: Liquid. In DMSO. Long Term Storage: -20°C.