Learn More
CPC Scientific (Z-Asp-Glu-Val-Asp)2-R110 1MG
SDP
Catalog No. 502109038
Encompass
Highly sensitive, photostable fluorogenic substrate for the determination of intracellular caspase-3 activity. The fluorophore rhodamine 110(Rh110) can be observed at an emission wavelength fo 535 nm with an excitation wavelength of 485 nm. With (H-Asp-Glu-Val-Asp)2-Rhodamine 110 (M-2620) the fluorescence intensity can be measured by flow cytometry, microwell plate reader, or fluorescence microscopy.SEQUENCE: (Z-Asp-Glu-Val-Asp)2-R110ONE-LETTER SEQUENCE: (Z-DEVD)2-R110MOLECULAR FORMULA: C72H78N10O27MOLECULAR WEIGHT: 1515.44STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [223538-61-2]SYNONYMS: (Z-Asp-Glu-Val-Asp)-Rhodamine 110RESEARCH AREA: ApoptosisCATEGORY: Apoptosis Related Peptides REFERENCES:Science 281, 1312 (1998)J. Biol. Chem., 276, 6974 (2001)Biochemistry 38, 13906 (1999)SKU(s): CASP-066A, CASP-066B, CASP-066C, CASP-066D
Catalog No. 50-210-9038 Supplier CPC Scientific Supplier No. CASP066B
Highly sensitive, photostable fluorogenic substrate for the determination of intracellular caspase-3 activity. The fluorophore rhodamine 110(Rh110) can be observed at an emission wavelength fo 535 nm with an excitation wavelength of 485 nm. With (H-Asp-Glu-Val-Asp)2-Rhodamine 110 (M-2620) the fluorescence intensity can be measured by flow cytometry, microwell plate reader, or fluorescence microscopy.SEQUENCE: (Z-Asp-Glu-Val-Asp)2-R110ONE-LETTER SEQUENCE: (Z-DEVD)2-R110MOLECULAR FORMULA: C72H78N10O27MOLECULAR WEIGHT: 1515.44STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [223538-61-2]SYNONYMS: (Z-Asp-Glu-Val-Asp)-Rhodamine 110RESEARCH AREA: ApoptosisCATEGORY: Apoptosis Related Peptides REFERENCES:Science 281, 1312 (1998)J. Biol. Chem., 276, 6974 (2001)Biochemistry 38, 13906 (1999)SKU(s): CASP-066A, CASP-066B, CASP-066C, CASP-066D