Learn More
Medchemexpress LLC Z-DL-Met-OH | 4434-61-1 | 283.34 | 25 G
SDP
Catalog No. 5000366630
Encompass_Preferred
Z-DL-Met-OH is a Methionine derivative. This solid, white to off-white compound is intended for research use only. Amino acids and their derivatives, like Z-DL-Met-OH, are utilized as ergogenic supplements. They play a role in influencing the secretion of anabolic hormones, supplying fuel during exercise, enhancing mental performance in stressful tasks, and preventing exercise-induced muscle damage.
  • Methionine derivative
  • Influences secretion of anabolic hormones
  • Supplies fuel during exercise
  • Enhances mental performance during stressful tasks
  • Prevents exercise-induced muscle damage
  • Recognized as a beneficial ergogenic dietary substance
  • Store at room temperature for 3 years
  • In solvent, store at -80°C for 2 years or -20°C for 1 year
Catalog No. 50-003-66630 Supplier Medchemexpress LLC Supplier No. 25GHYW009321
Z-DL-Met-OH is a Methionine derivative. This solid, white to off-white compound is intended for research use only. Amino acids and their derivatives, like Z-DL-Met-OH, are utilized as ergogenic supplements. They play a role in influencing the secretion of anabolic hormones, supplying fuel during exercise, enhancing mental performance in stressful tasks, and preventing exercise-induced muscle damage.
  • Methionine derivative
  • Influences secretion of anabolic hormones
  • Supplies fuel during exercise
  • Enhances mental performance during stressful tasks
  • Prevents exercise-induced muscle damage
  • Recognized as a beneficial ergogenic dietary substance
  • Store at room temperature for 3 years
  • In solvent, store at -80°C for 2 years or -20°C for 1 year