Learn More
Apexbio Technology LLC Z-DQMD-FMK 767287-99-0 5mg
SDP
Catalog No. 5000096916
Encompass_Preferred
Z-DQMD-FMK is a small-molecule inhibitor targeting caspase-3 It is designed to selectively and irreversibly inhibit caspase-3 thereby modulating caspase-3-mediated signaling pathways and apoptotic processes Z-DQMD-FMK exerts its biological activity primarily through covalent binding to active thiol residues of caspase-3 via its fluoromethyl ketone group resulting in irreversible inhibition Based on these pharmacological properties Z-DQMD-FMK holds research potential in investigating apoptotic mechanisms caspase-3-dependent signaling and proteolytic processing events including protein kinase C isoform cleavage under zinc deficiency oxidative stress and ischemic injury
Catalog No. 50-000-96916 Supplier Apexbio Technology LLC Supplier No. A19215
Z-DQMD-FMK is a small-molecule inhibitor targeting caspase-3 It is designed to selectively and irreversibly inhibit caspase-3 thereby modulating caspase-3-mediated signaling pathways and apoptotic processes Z-DQMD-FMK exerts its biological activity primarily through covalent binding to active thiol residues of caspase-3 via its fluoromethyl ketone group resulting in irreversible inhibition Based on these pharmacological properties Z-DQMD-FMK holds research potential in investigating apoptotic mechanisms caspase-3-dependent signaling and proteolytic processing events including protein kinase C isoform cleavage under zinc deficiency oxidative stress and ischemic injury