Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Z-Entacapone | 145195-63-7 | 305.29 | 25 MG
SDP

Catalog No. 5000349147
Encompass_Preferred

(Z)-Entacapone is a metabolite of the catechol-O-methyltransferase (COMT) inhibitor Entacapone. It is also a potential impurity found in commercial preparations of Entacapone and a degradant of Entacapone formed by UV light exposure.

  • Metabolite of the catechol-O-methyltransferase (COMT) inhibitor Entacapone
  • Potential impurity in commercial Entacapone preparations
  • Degradant of Entacapone formed by UV light exposure
  • Molecular formula: C14H15N3O5
  • SMILES: O=C(N(CC)CC)/C(C#N)=C\C1=CC([N+]([O-])=O)=C(O)C(O)=C1
  • Targets COMT
  • Involves metabolic enzyme/protease and neuronal signaling pathways

Catalog No. 50-003-49147 Supplier Medchemexpress LLC Supplier No. 25MGHY139089
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Z)-Entacapone is a metabolite of the catechol-O-methyltransferase (COMT) inhibitor Entacapone. It is also a potential impurity found in commercial preparations of Entacapone and a degradant of Entacapone formed by UV light exposure.

  • Metabolite of the catechol-O-methyltransferase (COMT) inhibitor Entacapone
  • Potential impurity in commercial Entacapone preparations
  • Degradant of Entacapone formed by UV light exposure
  • Molecular formula: C14H15N3O5
  • SMILES: O=C(N(CC)CC)/C(C#N)=C\C1=CC([N+]([O-])=O)=C(O)C(O)=C1
  • Targets COMT
  • Involves metabolic enzyme/protease and neuronal signaling pathways

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.