Learn More
Medchemexpress LLC (Z)-Entacapone | 145195-63-7 | MFCD00866580 | >98.0% | C14H15N3O5 | 10MG
SDP
Catalog No. 5000260609
Encompass_Preferred
(Z)-Entacapone is the Z-isomer of entacapone, a catechol-O-methyltransferase (COMT) inhibitor supplied for research and analytical use. It is commonly used as a reference or impurity standard and for studies of COMT activity, metabolism, and degradation pathways. The compound has molecular formula C14H15N3O5 and CAS 145195-63-7.
  • High chemical purity (>98.0%).
  • Suitable as a reference or impurity standard for analytical assays.
  • Applicable for studies of COMT activity, metabolism, and degradation.
  • Provided as a solid in small-quantity packs for research use.
  • Well characterized molecular identity (C14H15N3O5; CAS 145195-63-7).
Catalog No. 50-002-60609 Supplier Medchemexpress LLC Supplier No. HY13908910MG
(Z)-Entacapone is the Z-isomer of entacapone, a catechol-O-methyltransferase (COMT) inhibitor supplied for research and analytical use. It is commonly used as a reference or impurity standard and for studies of COMT activity, metabolism, and degradation pathways. The compound has molecular formula C14H15N3O5 and CAS 145195-63-7.
  • High chemical purity (>98.0%).
  • Suitable as a reference or impurity standard for analytical assays.
  • Applicable for studies of COMT activity, metabolism, and degradation.
  • Provided as a solid in small-quantity packs for research use.
  • Well characterized molecular identity (C14H15N3O5; CAS 145195-63-7).