Learn More
Apexbio Technology LLC Z-FA-FMK 105637-38-5;197855-65-5 10mg
SDP
Catalog No. 5000061640
Encompass_Preferred
Z-FA-FMK is a peptidyl fluoromethyl ketone inhibitor targeting cysteine proteases specifically cathepsins such as cathepsin B and certain caspases It does not require a P1 aspartate residue for inhibition Mechanistically Z-FA-FMK suppresses the enzyme activity of effector caspases (including caspases-2 -3 -6 and -7) but shows minimal impact on initiator caspases (caspase-8 and caspase-10) and only partial attenuation of caspase-9 activity As a negative control inhibitor Z-FA-FMK is regularly applied in apoptosis research to differentiate specific caspase-dependent signaling pathways and to assess the role of cathepsins and caspases in biological contexts such as cell death mechanisms and related pathways
Catalog No. 50-000-61640 Supplier Apexbio Technology LLC Supplier No. A817010
Z-FA-FMK is a peptidyl fluoromethyl ketone inhibitor targeting cysteine proteases specifically cathepsins such as cathepsin B and certain caspases It does not require a P1 aspartate residue for inhibition Mechanistically Z-FA-FMK suppresses the enzyme activity of effector caspases (including caspases-2 -3 -6 and -7) but shows minimal impact on initiator caspases (caspase-8 and caspase-10) and only partial attenuation of caspase-9 activity As a negative control inhibitor Z-FA-FMK is regularly applied in apoptosis research to differentiate specific caspase-dependent signaling pathways and to assess the role of cathepsins and caspases in biological contexts such as cell death mechanisms and related pathways