Learn More
Apexbio Technology LLC Z-FA-FMK, 1mg. Cas: 105637-38-5;197855-65-5 MFCD: MFCD00883668. Cysteine proteases inhibitor
SDP
Supplier:  Apexbio Technology LLC A81701
Encompass_Preferred
Z-FA-FMK is a control peptidic fluoromethylketone (boc-Thr-CH2F), and inhibitor to calpain (Ac-Leu-Leu-norleucinal), cathepsin B (Z-Phe-Ala-CH2F), and CPP32-like proteases (Z-DEVD-CH2F) [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5067