Learn More
Medchemexpress LLC HY-P0109A 1mg Medchemexpress, Z-FA-FMK CAS:197855-65-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0109A1MG
Encompass_Preferred
Medchemexpress, HY-P0109A 1mg Z-FA-FMK CAS:197855-65-5 Z-FA-FMK ((1S)-Z-FA-FMK; Compound 6) is a broad-spectrum halomethyl ketone inhibitor sgainst Coronavirus (SARS-CoV) main protease 3CL with a Ki of 25.7 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3020