Learn More
Abcam Z-FA-FMK, Cysteine protease inhibitor, 1MG
Catalog No. 502706578
Encompass_Preferred
MW 386.4 Da, Purity >95%. Irreversible cysteine protease inhibitor. Cell-permeable caspase effector inhibitor. Selectively inhibits recombinant effector caspases 2, -3, -6, and -7. Inhibits cathepsin B, L, and S, papain and cruzain. Induces apoptosis and necrosis. Shows anti-inflammatory effects in vivo. Negative control for caspase inhibitors. Liquid form also available (ab141384).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6578 Supplier Abcam Supplier No. AB1414841MG
MW 386.4 Da, Purity >95%. Irreversible cysteine protease inhibitor. Cell-permeable caspase effector inhibitor. Selectively inhibits recombinant effector caspases 2, -3, -6, and -7. Inhibits cathepsin B, L, and S, papain and cruzain. Induces apoptosis and necrosis. Shows anti-inflammatory effects in vivo. Negative control for caspase inhibitors. Liquid form also available (ab141384).
The product is subject to the following: Abcam Restricted Use Statement