Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Z)-FeCP-oxindole | 1137967-28-2 | 99.6% | 329.17 | 500 MG
SDP

Catalog No. 5000346912
Encompass_Preferred

(Z)-FeCP-oxindole is a selective human vascular endothelial growth factor receptor 2 (VEGFR2) inhibitor with an IC50 value of 200 nM. It can significantly inhibit VEGFR1 and PDGFRα or β at 10 μM. (Z)-FeCP-oxindole has some anticancer activity, acting on B16 murine melanoma lines with IC50 less than 1 μM.

  • Target: VEGFR
  • Pathway: Protein Tyrosine Kinase/RTK
  • Storage: Powder: -20°C for 3 years, 4°C for 2 years; in solvent: -80°C for 6 months, -20°C for 1 month
  • In vitro solubility: DMSO: 50 mg/mL (151.90 mM; need ultrasonic)
  • IC50 & target: VEGFR2 (200 nM), VEGFR1

Catalog No. 50-003-46912 Supplier Medchemexpress LLC Supplier No. HY10844450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Z)-FeCP-oxindole is a selective human vascular endothelial growth factor receptor 2 (VEGFR2) inhibitor with an IC50 value of 200 nM. It can significantly inhibit VEGFR1 and PDGFRα or β at 10 μM. (Z)-FeCP-oxindole has some anticancer activity, acting on B16 murine melanoma lines with IC50 less than 1 μM.

  • Target: VEGFR
  • Pathway: Protein Tyrosine Kinase/RTK
  • Storage: Powder: -20°C for 3 years, 4°C for 2 years; in solvent: -80°C for 6 months, -20°C for 1 month
  • In vitro solubility: DMSO: 50 mg/mL (151.90 mM; need ultrasonic)
  • IC50 & target: VEGFR2 (200 nM), VEGFR1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.