Learn More
Medchemexpress LLC (Z)-FeCP-oxindole | 1137967-28-2 | 99.62% | 329.17 | 25 MG
SDP
Catalog No. 5000347542
Encompass_Preferred
(Z)-FeCP-oxindole is a selective human vascular endothelial growth factor receptor 2 (VEGFR2) inhibitor with an IC50 value of 200 nM. It can significantly inhibit VEGFR1 and PDGFRα or β at 10 μM. (Z)-FeCP-oxindole also exhibits anticancer activity, specifically acting on B16 murine melanoma lines with an IC50 less than 1 μM.
  • Selective human vascular endothelial growth factor receptor 2 (VEGFR2) inhibitor.
  • Inhibits VEGFR1 and PDGFRα or β.
  • Has anticancer activity on B16 murine melanoma lines.
  • For research use only.
Catalog No. 50-003-47542 Supplier Medchemexpress LLC Supplier No. 25MGHY108444
(Z)-FeCP-oxindole is a selective human vascular endothelial growth factor receptor 2 (VEGFR2) inhibitor with an IC50 value of 200 nM. It can significantly inhibit VEGFR1 and PDGFRα or β at 10 μM. (Z)-FeCP-oxindole also exhibits anticancer activity, specifically acting on B16 murine melanoma lines with an IC50 less than 1 μM.
  • Selective human vascular endothelial growth factor receptor 2 (VEGFR2) inhibitor.
  • Inhibits VEGFR1 and PDGFRα or β.
  • Has anticancer activity on B16 murine melanoma lines.
  • For research use only.