Learn More
TARGETMOL CHEMICALS INC Z-FECP-OXINDOLE 5MG
SDP
Catalog No. 502580859
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (Z)-FeCP-oxindole is a selective inhibitor of VEGFR2 with an IC50 of 200 nM for human. (Z)-FeCP-oxindole shows anticancer activity. purity: 98%
Catalog No. 50-258-0859 Supplier TARGETMOL CHEMICALS INC Supplier No. T235545MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (Z)-FeCP-oxindole is a selective inhibitor of VEGFR2 with an IC50 of 200 nM for human. (Z)-FeCP-oxindole shows anticancer activity. purity: 98%