Learn More
Medchemexpress LLC Z-FF-FMK | 105608-85-3 | 96.0% | C27H27FN2O4 | 25 MG
SDP
Catalog No. 5000352505
Encompass_Preferred
Z-FF-FMK is a selective cathepsin-L inhibitor. It can prevent β-amyloid from inducing apoptotic changes such as activation of caspase-3, cleavage of the DNA repair enzyme, poly-ADP ribose polymerase, and DNA fragmentation.
  • Selective cathepsin-L inhibitor
  • Prevents β-amyloid induced apoptotic changes
  • Inhibits caspase-3 activation
  • Cleavage of DNA repair enzyme, poly-ADP ribose polymerase, and DNA fragmentation
Catalog No. 50-003-52505 Supplier Medchemexpress LLC Supplier No. 25MGHY141867
Z-FF-FMK is a selective cathepsin-L inhibitor. It can prevent β-amyloid from inducing apoptotic changes such as activation of caspase-3, cleavage of the DNA repair enzyme, poly-ADP ribose polymerase, and DNA fragmentation.
  • Selective cathepsin-L inhibitor
  • Prevents β-amyloid induced apoptotic changes
  • Inhibits caspase-3 activation
  • Cleavage of DNA repair enzyme, poly-ADP ribose polymerase, and DNA fragmentation