Learn More
Medchemexpress LLC Z-fg-nho-bzome | 180313-86-4 | 98.0% | 505.52 | C27H27N3O7 | 10mg
SDP
Catalog No. 5000269014
Encompass_Preferred
Z-FG-NHO-BzOME is a cysteine protease inhibitor that selectively inhibits cathepsin B cathepsin L cathepsin S and papain[1]
Catalog No. 50-002-69014 Supplier Medchemexpress LLC Supplier No. HYW79001410MG
Z-FG-NHO-BzOME is a cysteine protease inhibitor that selectively inhibits cathepsin B cathepsin L cathepsin S and papain[1]