Learn More
Anatech Ltd Z-FIX/4/GALLON
Catalog No. NC9937162
Encompass_Preferred
Z-FIX,,INGREDIENTS:Formaldehyde - 3.7 %; Ionized zinc; Buffer,PHYSICAL AND CHEMICAL DATA:Boiling point:>212deg.F;Evaporation rate (n butyl acetate=1):<1;Vapor density (air=1): 1; Solubility in water:Complete ; Specific
Catalog No. NC9937162 Supplier Anatech Ltd Supplier No. 174
Z-FIX,,INGREDIENTS:Formaldehyde - 3.7 %; Ionized zinc; Buffer,PHYSICAL AND CHEMICAL DATA:Boiling point:>212deg.F;Evaporation rate (n butyl acetate=1):<1;Vapor density (air=1): 1; Solubility in water:Complete ; Specific