Learn More
Anatech Ltd Z-FIX ZINC FIXATIVE 5 GAL
Catalog No. NC9378601
Encompass_Preferred
Fixative, Buffered Zinc Formalin; Anatech; Ready-to-Use; No formalin figment; Does not precipitate in processors or tissues; 5 gal.
Catalog No. NC9378601 Supplier Anatech Ltd Supplier No. 175
Fixative, Buffered Zinc Formalin; Anatech; Ready-to-Use; No formalin figment; Does not precipitate in processors or tissues; 5 gal.