Learn More
Enzo Life Sciences OMNICATHEPSIN fluorogenic substrate (50mg). CAS: 65147-22-0
SDP
Catalog No. 502011251
Encompass_Preferred
Fluorogenic substrate cleaved by plasma kallikrein and kallikrein 8, but not plasmin; by papain; and by most cathepsins (cathepsins B, C/DPP-I, F, K/O2, L, L2/V, O, S, and X/Z). Ex.: 380 nm, Em.: 460 nm, although the following Ex/Em can also be used: 355,375/440,450. This substrate is useful for inhibitor screening and kinetic analysis. Also available: fluorogenic calibration standard AMC (BML-KI144). Alternative name: Z-FR-AMC, Z-Phe-Arg-AMC. Purity: ≥99%. Long Term Storage: -20°C.
Catalog No. 50-201-1251 Supplier Enzo Life Sciences Supplier No. BMLP1390050
Fluorogenic substrate cleaved by plasma kallikrein and kallikrein 8, but not plasmin; by papain; and by most cathepsins (cathepsins B, C/DPP-I, F, K/O2, L, L2/V, O, S, and X/Z). Ex.: 380 nm, Em.: 460 nm, although the following Ex/Em can also be used: 355,375/440,450. This substrate is useful for inhibitor screening and kinetic analysis. Also available: fluorogenic calibration standard AMC (BML-KI144). Alternative name: Z-FR-AMC, Z-Phe-Arg-AMC. Purity: ≥99%. Long Term Storage: -20°C.