Learn More
Medchemexpress LLC HY-128140 5mg , Z-FY-CHO CAS:167498-29-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1281405MG
Encompass_Preferred
HY-128140 5mg Z-FY-CHO CAS: 167498-29-5 Z-FY-CHO (Z-Phe-Tyr-CHO) is a potent and specific cathepsin L (CTSL) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0016