Learn More
Bachem Z-Gly-Gly-Arg-bNA · HCl
SDP
Supplier:  Bachem 4000903.1000
Encompass_Preferred
Z-Gly-Gly-Arg-bNA · HCl, 1442-79-1, C28H33N7O5 · HCl, Mr 584.08, 1g. Z-GGR-bNA, sensitive chromogenic substrate for trypsin. Furthermore, this substrate allows a distinct histochemical localization of enteropeptidase with the trypsinogen method. It is also cleaved by neutral endopeptidase 24.5. Z-GGR-bNA is also used for determining the tryptic activity of the 20S proteasome.
Catalog No. 50-194-7035