Learn More
Carbosynth LLC. Z-Gln-Gly, 1g, MW: 337.33, Molecular formula: C15H19N3O6, CAS# 6610-42-0
SDP
Catalog No. 501687182
Encompass_Preferred
Synonyms: Benzyloxycarbonyl-L-Glutaminyl-Glycine, Purity: , Grade AA, Application: Substrate for Transglutaminase, Storage Temperature: -20°C, References: J.E. Folk and P.W. Cole, J. Biol. Chem., 241, 5518 (1966)., . H. Ando, M. Adachi, K. Umeda, A. Matsuura, M. Nonaka, R. Uchio, H. Tanaka, and M. Motoki, Agric. Biol. Chem., 53, 2613 (1989).
Catalog No. 50-168-7182 Supplier Carbosynth LLC. Supplier No. SZQ31901G
Synonyms: Benzyloxycarbonyl-L-Glutaminyl-Glycine, Purity: , Grade AA, Application: Substrate for Transglutaminase, Storage Temperature: -20°C, References: J.E. Folk and P.W. Cole, J. Biol. Chem., 241, 5518 (1966)., . H. Ando, M. Adachi, K. Umeda, A. Matsuura, M. Nonaka, R. Uchio, H. Tanaka, and M. Motoki, Agric. Biol. Chem., 53, 2613 (1989).