Learn More
Bachem Z-Gln-Gly-OH
SDP
Catalog No. NC1726909
Encompass_Preferred
Z-Gln-Gly-OH, 6610-42-0, C15H19N3O6, Mr 337.33, 5g. Substrate for tissue transaminase. De Macédo et al. described a direct continuous spectrophotometric assay for transglutaminase (TGase) activity.
Catalog No. NC1726909 Supplier Bachem Supplier No. 4002246.0005
Add to Cart
Add to Cart
Z-Gln-Gly-OH, 6610-42-0, C15H19N3O6, Mr 337.33, 5g. Substrate for tissue transaminase. De Macédo et al. described a direct continuous spectrophotometric assay for transglutaminase (TGase) activity.